Learn More
Abnova™ Human NT5DC2 Full-length ORF (NP_075059.1, 1 a.a. - 520 a.a.) Recombinant Protein with GST-tag at N-terminal
Recombinant proteins expressed from in vitro wheat germ system.
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_075059.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Gene ID (Entrez) | 64943 |
Name | NT5DC2 (Human) Recombinant Protein (P01) |
Purification Method | Glutathione Sepharose 4 Fast Flow |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16193359
|
Abnova™
H00064943-P01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16103369
|
Abnova™
H00064943-P01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Good Folding
- Excellent amino acids labeling
Immunogen Sequence: MRVESGSAQERGILLESLSTLLEKTTASHEGRAPGNRELTDLLPPEVCSLLNPAAIYANNEISLRDVEVYGFDYDYTLAQYADALHPEIFSTARDILIEHYKYPEGIRKYDYNPSFAIRGLHYDIQKSLLMKIDAFHYVQLGTAYR
Specifications
NP_075059.1 | |
64943 | |
Glutathione Sepharose 4 Fast Flow | |
MRVESGSAQERGILLESLSTLLEKTTASHEGRAPGNRELTDLLPPEVCSLLNPAAIYANNEISLRDVEVYGFDYDYTLAQYADALHPEIFSTARDILIEHYKYPEGIRKYDYNPSFAIRGLHYDIQKSLLMKIDAFHYVQLGTAYRGLQPVPDEEVIELYGGTQHIPLYQMSGFYGKGPSIKQFMDIFSLPEMALLSCVVDYFLGHSLEFDQAHLYKDVTDAIRDVHVKGLMYQWIEQDMEKYILRGDETFAVLSRLVAHGKQLFLITNSPFSFVDKGMRHMVGPDWRQLFDVVIVQADKPSFFTDRRKPFRKLDEKGSLQWDRITRLEKGKIYRQGNLFDFLRLTEWRGPRVLYFGDHLYSDLADLMLRHGWRTGAIIPELEREIRIINTEQYMHSLTWQQALTGLLERMQTYQDAESRQVLAAWMKERQELRCITKALFNAQFGSIFRTFHNPTYFSRRLVRFSDLYMASLSCLLNYRVDFTFYPRRTPLQHEAPLWMDQLCTGCMKTPFLGDMAHIR | |
RUO | |
NT5DC2 | |
GST |
Antibody Production, Protein Array, ELISA, Western Blot | |
NT5DC2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ12442 | |
NT5DC2 | |
Liquid |
For Research Use Only