Learn More
Abnova™ Human NSDHL Full-length ORF (NP_057006.1, 1 a.a. - 373 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00050814-P01.10ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is localized in the endoplasmic reticulum and is involved in cholesterol biosynthesis. Mutations in this gene are associated with CHILD syndrome, which is a X-linked dominant disorder of lipid metabolism with disturbed cholesterol biosynthesis, and typically lethal in males. Alternatively spliced transcript variants with differing 5' UTR have been found for this gene. [provided by RefSeq]
Sequence: MEPAVSEPMRDQVARTHLTEDTPKVNADIEKVNQNQAKRCTVIGGSGFLGQHMVEQLLARGYAVNVFDIQQGFDNPQVRFFLGDLCSRQDLYPALKGVNTVFHCASPPPSSNNKELFYRVNYIGTKNVIETCKEAGVQKLILTSSASVIFEGVDIKNGTEDLPYAMKPIDYYTETKILQERAVLGANDPEKNFLTTAIRPHGIFGPRDPQLVPILIEAARNGKMKFVIGNGKNLVDFTFVENVVHGHILAAEQLSRDSTLGGKAFHITNDEPIPFWTFLSRILTGLNYEAPKYHIPYWVAYYLALLLSLLVMVISPVIQLQPTFTPMRVALAGTFHYYSCERAKKAMGYQPLVTMDDAMERTVQSFRHLRRVKSpecifications
NP_057006.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
68.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
H105E3/SDR31E1/XAP104 | |
NSDHL | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
50814 | |
NSDHL (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MEPAVSEPMRDQVARTHLTEDTPKVNADIEKVNQNQAKRCTVIGGSGFLGQHMVEQLLARGYAVNVFDIQQGFDNPQVRFFLGDLCSRQDLYPALKGVNTVFHCASPPPSSNNKELFYRVNYIGTKNVIETCKEAGVQKLILTSSASVIFEGVDIKNGTEDLPYAMKPIDYYTETKILQERAVLGANDPEKNFLTTAIRPHGIFGPRDPQLVPILIEAARNGKMKFVIGNGKNLVDFTFVENVVHGHILAAEQLSRDSTLGGKAFHITNDEPIPFWTFLSRILTGLNYEAPKYHIPYWVAYYLALLLSLLVMVISPVIQLQPTFTPMRVALAGTFHYYSCERAKKAMGYQPLVTMDDAMERTVQSFRHLRRVK | |
RUO | |
NSDHL | |
Wheat Germ (in vitro) | |
GST | |
Liquid |