Learn More
Abnova™ Human NRG2 Partial ORF (NP_004874, 116 a.a. - 215 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009542-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Neuregulin 2 (NRG2) is a novel member of the neuregulin family of growth and differentiation factors. Through interaction with the Erbb family of receptors, NRG2 induces the growth and differentiation of epithelial, neuronal, glial, and other types of cells. The gene consists of 12 exons and the genomic structure is similar to that of neuregulin 1 (NRG1), another member of the neuregulin family of ligands. NRG1 and NRG2 mediate distinct biological processes by acting at different sites in tissues and eliciting different biological responses in cells. The gene is located close to the region for demyelinating Charcot-Marie-Tooth disease locus, but is not responsible for this disease. Alternative transcripts encoding distinct isoforms have been described. [provided by RefSeq]
Sequence: SLKSVQDQAYKAPVVVEGKVQGLVPAGGSSSNSTREPPASGRVALVKVLDKWPLRSGGLQREQVISVGSCVPLERNQRYIFFLEPTEQPLVFKTAFAPLDSpecifications
NP_004874 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SLKSVQDQAYKAPVVVEGKVQGLVPAGGSSSNSTREPPASGRVALVKVLDKWPLRSGGLQREQVISVGSCVPLERNQRYIFFLEPTEQPLVFKTAFAPLD | |
RUO | |
NRG2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
9542 | |
NRG2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
Don-1/HRG2/NTAK | |
NRG2 | |
Recombinant | |
wheat germ expression system |