missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human NREP Partial ORF (NP_004763.1, 1 a.a. - 68 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00009315-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16140356

  • 3602.46 NOK / 10µg

Estimated Shipment: 21-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: MVYYPELFVWVSQEPFPNKDMEGRLPKGRLPVPKEVNRKKNDETNAASLTPLGSSELRSPRISYLHFF
Specifications

Specifications

NP_004763.1
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
33.22kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MVYYPELFVWVSQEPFPNKDMEGRLPKGRLPVPKEVNRKKNDETNAASLTPLGSSELRSPRISYLHFF
RUO
NREP
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
9315
NREP (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C5orf13/D4S114/P311/PRO1873/PTZ17/SEZ17
NREP
Yes
Videos
SDS
Documents
Certificates