missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human NRBP Partial ORF (AAH01221, 436 a.a. - 535 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00029959-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16186196

  • 4115.00 NOK / 10µg
Estimated Shipment: 02-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: PAEVETRKVVLMQCNIESVEEGVKHHLTLLLKLEDKLNRHLSCDLMPNENIPELAAELVQLGFISEADQSRLTSLLEETLNKFNFARNSTLNSAAVTVSS
Specifications

Specifications

AAH01221
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
PAEVETRKVVLMQCNIESVEEGVKHHLTLLLKLEDKLNRHLSCDLMPNENIPELAAELVQLGFISEADQSRLTSLLEETLNKFNFARNSTLNSAAVTVSS
RUO
NRBP1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
29959
NRBP (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BCON3/FLJ27109/FLJ35541/MADM/MUDPNP/NRBP
NRBP1
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers