missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human NRAP Partial ORF (NP_932326.2, 1640 a.a. - 1727 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00004892-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16122075

  • 4115.00 NOK / 10µg
Estimated Shipment: 01-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: LRHAQKAHQLQSDVKYKSDLNLTRGVGWTPPGSYKVEMARRAAELANARGLGLQGAYRGAEAVEAGDHQSGEVNPDATEILHVKKKKA
Specifications

Specifications

NP_932326.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.42kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
LRHAQKAHQLQSDVKYKSDLNLTRGVGWTPPGSYKVEMARRAAELANARGLGLQGAYRGAEAVEAGDHQSGEVNPDATEILHVKKKKA
RUO
NRAP
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
4892
NRAP (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
NRAP
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers