missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human NR2C2 Partial ORF (NP_003289, 43 a.a. - 152 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00007182-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16146935

  • 6245.00 NOK / 25µg
Estimated Shipment: 01-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Members of the nuclear hormone receptor family, such as NR2C2, act as ligand-activated transcription factors. The proteins have an N-terminal transactivation domain, a central DNA-binding domain with 2 zinc fingers, and a ligand-binding domain at the C terminus. The activated receptor/ligand complex is translocated to the nucleus where it binds to hormone response elements of target genes (Yoshikawa et al., 1996 [PubMed 8661150]).[supplied by OMIM]

Sequence: KIVTDQQTGQKIQIVTAVDASGSPKQQFILTSPDGAGTGKVILASPETSSAKQLIFTTSDNLVPGRIQIVTDSASVERLLGKTDVQRPQVVEYCVVCGDKASGRHYGAVS
Specifications

Specifications

NP_003289
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
KIVTDQQTGQKIQIVTAVDASGSPKQQFILTSPDGAGTGKVILASPETSSAKQLIFTTSDNLVPGRIQIVTDSASVERLLGKTDVQRPQVVEYCVVCGDKASGRHYGAVS
RUO
NR2C2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
7182
NR2C2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
TAK1/TR2R1/TR4/hTAK1
NR2C2
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers