missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human NLRP2 Full-length ORF (NP_060322.1, 1 a.a. - 1062 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00055655-P02.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16111643

  • 4115.00 NOK / 10µg
Estimated Shipment: 02-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

NALP proteins, such as NALP2, are characterized by an N-terminal pyrin (MIM 608107) domain (PYD) and are involved in the activation of caspase-1 (CASP1; MIM 147678) by Toll-like receptors (see TLR4; MIM 603030). They may also be involved in protein complexes that activate proinflammatory caspases (Tschopp et al., 2003 [PubMed 12563287]).[supplied by OMIM]

Sequence: MVSSAQMGFNLQALLEQLSQDELSKFKYLITTFSLAHELQKIPHKEVDKADGKQLVEILTTHCDSYWVEMASLQVFEKMHRMDLSERAKDEVREAALKSFNKRKPLSLGITRKERPPLDVDEMLERFKTEAQAFTETKGNVICLGKEVFKGKKPDKDNRCRYILKTKFREMWKSWPGDSKEVQVMAERYKMLIPFSNPRVLPGPFSYTVVLYGPAGLGKTTLAQKLMLDWAEDNLIHKFKYAFYLSCRELSRLGPCSFAELVFRDWPELQDDIPHILAQARKILFVIDGFDELGAAPGALIEDICGDWEKKKPVPVLLGSLLNRVMLPKAALLVTTRPRALRDLRILAEEPIYIRVEGFLEEDRRAYFLRHFGDEDQAMRAFELMRSNAALFQLGSAPAVCWIVCTTLKLQMEKGEDPVPTCLTRTGLFLRFLCSRFPQGAQLRGALRTLSLLAAQGLWAQTSVLHREDLERLGVQESDLRLFLDGDILRQDRVSKGCYSFIHLSFQQFLTALFYTLEKEEEEDRDGHTWDIGDVQKLLSGVERLRNPDLIQAGYYSFGLANEKRAKELEATFGCRMSPDIKQELLRCDISCKGGHSTVTDLQELLGCLYESQEEELVKEVMAQFKEISLHLNAVDVVPSSFCVKHCRNLQKMSLQVIKENLPENVTASESDAEVERSQDDQHMLPFWTDLCSIFGSNKDLMGLAINDSFLSASLVRILCEQIASDTCHLQRVVFKNISPADAHRNLCLALRGHKTVTYLTLQGNDQDDMFPALCEVLRHPECNLRYLGLVSCSATTQQWADLSLALEVNQSLTCVNLSDNELLDEGAKLLYTTLRHPKCFLQRLSLENCHLTEANCKDLAAVLVVSRELTHLCLAKNPIGNTGVKFLCEGLRYPECKLQTLVLWNCDITSDGCCDLTKLLQEKSSLLCLDLGLNHIGVKGMKFLCEALRKPLCNLRCLWLWGCSIPPFSCEDLCSALSCNQSLVTLDLGQNPLGSSGVKMLFETLTCSSGTLRTLRLKIDDFNDELNKLLEEIEEKNPQLIIDTEKHHPWAERPSSHDFMI
Specifications

Specifications

NP_060322.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
146.9kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CLR19.9/FLJ20510/NALP2/NBS1/PAN1/PYPAF2
NLRP2
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
55655
NLRP2 (Human) Recombinant Protein (P02)
12.5% SDS-PAGE Stained with Coomassie Blue.
MVSSAQMGFNLQALLEQLSQDELSKFKYLITTFSLAHELQKIPHKEVDKADGKQLVEILTTHCDSYWVEMASLQVFEKMHRMDLSERAKDEVREAALKSFNKRKPLSLGITRKERPPLDVDEMLERFKTEAQAFTETKGNVICLGKEVFKGKKPDKDNRCRYILKTKFREMWKSWPGDSKEVQVMAERYKMLIPFSNPRVLPGPFSYTVVLYGPAGLGKTTLAQKLMLDWAEDNLIHKFKYAFYLSCRELSRLGPCSFAELVFRDWPELQDDIPHILAQARKILFVIDGFDELGAAPGALIEDICGDWEKKKPVPVLLGSLLNRVMLPKAALLVTTRPRALRDLRILAEEPIYIRVEGFLEEDRRAYFLRHFGDEDQAMRAFELMRSNAALFQLGSAPAVCWIVCTTLKLQMEKGEDPVPTCLTRTGLFLRFLCSRFPQGAQLRGALRTLSLLAAQGLWAQTSVLHREDLERLGVQESDLRLFLDGDILRQDRVSKGCYSFIHLSFQQFLTALFYTLEKEEEEDRDGHTWDIGDVQKLLSGVERLRNPDLIQAGYYSFGLANEKRAKELEATFGCRMSPDIKQELLRCDISCKGGHSTVTDLQELLGCLYESQEEELVKEVMAQFKEISLHLNAVDVVPSSFCVKHCRNLQKMSLQVIKENLPENVTASESDAEVERSQDDQHMLPFWTDLCSIFGSNKDLMGLAINDSFLSASLVRILCEQIASDTCHLQRVVFKNISPADAHRNLCLALRGHKTVTYLTLQGNDQDDMFPALCEVLRHPECNLRYLGLVSCSATTQQWADLSLALEVNQSLTCVNLSDNELLDEGAKLLYTTLRHPKCFLQRLSLENCHLTEANCKDLAAVLVVSRELTHLCLAKNPIGNTGVKFLCEGLRYPECKLQTLVLWNCDITSDGCCDLTKLLQEKSSLLCLDLGLNHIGVKGMKFLCEALRKPLCNLRCLWLWGCSIPPFSCEDLCSALSCNQSLVTLDLGQNPLGSSGVK
RUO
NLRP2
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers