Learn More
Abnova™ Human NLGN3 Full-length ORF (AAH51715.1, 1 a.a. - 828 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00054413-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of a family of neuronal cell surface proteins. Members of this family may act as splice site-specific ligands for beta-neurexins and may be involved in the formation and remodeling of central nervous system synapses. Mutations in this gene may be associated with autism and Asperger syndrome. Multiple transcript variants encoding distinct isoforms have been identified for this gene, but their full length sequences have not been determined. [provided by RefSeq]
Sequence: MWLRLGPPSLSLSPKPTVGRSLCLTLWFLSLALRASTQAPAPTVNTHFGKLRGARVPLPSEILGPVDQYLGVPYAAPPIGEKRFLPPEPPPYWSGIRNATHFPPVCPQNIHTAVPEVMLPVWFTANLDIVATYIQEPNEDCLYLNVYVPTEDGSGAKKQGEDLADNDGDEDEDIRDSGAKPVMVYIHGGSYMEGTGNMIDGSILASYGNVIVITLNYRVGVLGFLSTGDQAAKGNYGLLDQIQALRWVSENIAFFGGDPRRITVFGSGIGASCVSLLTLSHHSEGLFQRAIIQSGSALSSWAVNYQPVKYTSLLADKVGCNVLDTVDMVDCLRQKSAKELVEQDIQPARYHVAFGPVIDGDVIPDDPEILMEQGEFLNYDIMLGVNQGEGLKFVEGVVDPEDGVSGTDFDYSVSNFVDNLYGYPEGKDTLRETIKFMYTDWADRDNPETRRKTLVALFTDHQWVEPSVVTADLHARYGSPTYFYAFYHHCQSLMKPAWSDAAHGDEVPYVFGVPMVGPTDLFPCNFSKNDVMLSAVVMTYWTNFAKTGDPNKPVPQDTKFIHTKANRFEEVAWSKYNPRDQLYLHIGLKPRVRDHYRATKVAFWKHLVPHLYNLHDMFHYTSTTTKVPPPDTTHSSHITRRPNGKTWSTKRPAISPAYSNENAQGSWNGDQDAGPLLVENPRDYSTELSVTIAVGASILFLNVLAFAALYYRKDKRRQEPLRQPSPQRGAWAPELGAAPEEELAALQLGPTHHECEASPPHDTLRLTALPDYTLTLRRSPDDIPLMTPNTITMIPNSLVGLQTLHPYNTFAAGFNSTGLPHSHSTTRVSpecifications
AAH51715.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
118.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ASPGX1/AUTSX1/HNL3/KIAA1480 | |
NLGN3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
54413 | |
NLGN3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MWLRLGPPSLSLSPKPTVGRSLCLTLWFLSLALRASTQAPAPTVNTHFGKLRGARVPLPSEILGPVDQYLGVPYAAPPIGEKRFLPPEPPPYWSGIRNATHFPPVCPQNIHTAVPEVMLPVWFTANLDIVATYIQEPNEDCLYLNVYVPTEDGSGAKKQGEDLADNDGDEDEDIRDSGAKPVMVYIHGGSYMEGTGNMIDGSILASYGNVIVITLNYRVGVLGFLSTGDQAAKGNYGLLDQIQALRWVSENIAFFGGDPRRITVFGSGIGASCVSLLTLSHHSEGLFQRAIIQSGSALSSWAVNYQPVKYTSLLADKVGCNVLDTVDMVDCLRQKSAKELVEQDIQPARYHVAFGPVIDGDVIPDDPEILMEQGEFLNYDIMLGVNQGEGLKFVEGVVDPEDGVSGTDFDYSVSNFVDNLYGYPEGKDTLRETIKFMYTDWADRDNPETRRKTLVALFTDHQWVEPSVVTADLHARYGSPTYFYAFYHHCQSLMKPAWSDAAHGDEVPYVFGVPMVGPTDLFPCNFSKNDVMLSAVVMTYWTNFAKTGDPNKPVPQDTKFIHTKANRFEEVAWSKYNPRDQLYLHIGLKPRVRDHYRATKVAFWKHLVPHLYNLHDMFHYTSTTTKVPPPDTTHSSHITRRPNGKTWSTKRPAISPAYSNENAQGSWNGDQDAGPLLVENPRDYSTELSVTIAVGASILFLNVLAFAALYYRKDKRRQEPLRQPSPQRGAWAPELGAAPEEELAALQLGPTHHECEASPPHDTLRLTALPDYTLTLRRSPDDIPLMTPNTITMIPNSLVGLQTLHPYNTFAAGFNSTGLPHSHSTTRV | |
RUO | |
NLGN3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |