missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Nkx3.2 Synthetic Peptide

Product Code. 15295219
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15295219 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15295219 Supplier Invitrogen™ Supplier No. PEP1222

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Synthetic Peptide

BAPX1 Synthetic Peptide, PEP-1222, from Invitrogen.

This gene encodes a member of the NK family of homeobox-containing proteins. The encoded protein may play a role in skeletal development.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 579
Species Human
Purification Method purified
Accession Number P78367
Common Name Human Nkx3.2
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Symbol NKX3-2
Lyophilized No
Preservative 0.02% sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.