Learn More
Abnova™ Human NKTR Partial ORF (NP_005376.2, 888 a.a. - 984 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00004820-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a membrane-anchored protein with a hydrophobic amino terminal domain and a cyclophilin-like PPIase domain. It is present on the surface of natural killer cells and facilitates their binding to targets. Its expression is regulated by IL2 activation of the cells. [provided by RefSeq]
Sequence: ESNSERDVTKNSKNDSHPSSDKEEGEATSDSESEVSEIHIKVKPTTKSSTNTSLPDDNGAWKSSKQRTSTSDSEGSCSNSENNRGKPQKHKHGSKENSpecifications
NP_005376.2 | |
Liquid | |
4820 | |
NKTR (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686F1754/DKFZp686G0426/DKFZp686J06106/DKFZp686N24126/MGC90527/p104 | |
NKTR | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.41kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
ESNSERDVTKNSKNDSHPSSDKEEGEATSDSESEVSEIHIKVKPTTKSSTNTSLPDDNGAWKSSKQRTSTSDSEGSCSNSENNRGKPQKHKHGSKEN | |
RUO | |
NKTR | |
Wheat Germ (in vitro) | |
GST |