Learn More
Abnova™ Human NIPBL Full-length ORF (AAH33847.1, 1 a.a. - 175 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00025836-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes the homolog of the Drosophila melanogaster Nipped-B gene product and fungal Scc2-type sister chromatid cohesion proteins. The Drosophila protein facilitates enhancer-promoter communication of remote enhancers and plays a role in developmental regulation. It is also homologous to a family of chromosomal adherins with broad roles in sister chromatid cohesion, chromosome condensation, and DNA repair. The human protein has a bipartite nuclear targeting sequence and a putative HEAT repeat. Condensins, cohesins and other complexes with chromosome-related functions also contain HEAT repeats. Mutations in this gene result in Cornelia de Lange syndrome, a disorder characterized by dysmorphic facial features, growth delay, limb reduction defects, and mental retardation. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: MKCLPENSAPLIEFANVSQGILLLLMLKQHLKNLCGFSDSKIQKYSPSESAKVYDKAINRKTGVHFHPKQTLDFLRSDMANSKITEEVKRSIVKQYLDFKLLMEHLDPDEEEEEGEVSASTNARNKAITSLLGGGSPKNNTAAETEDDESDGEDRGGGTSGVRRRRSQRISQRITSpecifications
AAH33847.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
45.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CDLS/CDLS1/DKFZp434L1319/FLJ11203/FLJ12597/FLJ13354/FLJ13648/FLJ44854/IDN3/IDN3-B | |
NIPBL | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
25836 | |
NIPBL (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MKCLPENSAPLIEFANVSQGILLLLMLKQHLKNLCGFSDSKIQKYSPSESAKVYDKAINRKTGVHFHPKQTLDFLRSDMANSKITEEVKRSIVKQYLDFKLLMEHLDPDEEEEEGEVSASTNARNKAITSLLGGGSPKNNTAAETEDDESDGEDRGGGTSGVRRRRSQRISQRIT | |
RUO | |
NIPBL | |
Wheat Germ (in vitro) | |
GST | |
Liquid |