missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human NID2 Full-length ORF (AAH35608.1, 1 a.a. - 969 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | AAH35608.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 22795 |
Molecular Weight (g/mol) | 133.5kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16116592
|
Abnova™
H00022795-P01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 03-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16126592
|
Abnova™
H00022795-P01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 03-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Basement membranes, which are composed of type IV collagens (see MIM 120130), laminins (see LAMC1; MIM 150290), perlecan (HSPG2; MIM 142461), and nidogen (see NID1; MIM 131390), are thin pericellular protein matrices that control a large number of cellular activities, including adhesion, migration, differentiation, gene expression, and apoptosis.[supplied by OMIM]
Sequence: MEGDRVAGRPVLSSLPVLLLLQLLMLRAAALHPDELFPHGESWGDQLLQEGDDESSAVVKLANPLHFYEARFSNLYVGTNGIISTQDFPRETQYVDYDFPTDFPAIAPFLADIDTSHGRGRVLYRVNGKVSGHLHVGHTPVHFTDVDLHAYIVGNDGRAYTAISHIPQPAAQALLPLTPIGGLFGWLFALEKPGSENGFSLAGAAFTHDMEVTFYPGEETVRITQTAEGLDPENYLSIKTNIQGQVPYVPANFTAHISPYKELYHYSDSTVTSTSSRDYSLTFGAINQTWSYRIHQNITYQVCRHAPRHPSFPTTQQLNVDRVFALYNDEERVLRFAVTNQIGPVKEDSDPTPVNPCYDGSHMCDTTARCHPGTGVDYTCECASGYQGDGRNCVDENECATGFHRCGPNSVCINLPGSYRCECRSAYEFADDRHTCILITPPANPCEDGSHTCAPAGQARCVHHGGSTFSCACLPGYAGDGHQCTDVDECSENRCHPAATCYNTPGSFSCRCQPGYYGDGFQCIPDSTSSLTPCEQQQRHAQAQYAYPGARFHIPQCDEQGNFLPLQCHGSTGFCWCVDPDGHEVPGTQTPPGSTPPHCGPSPEPTQRPPTICERWRENLLEHYGGTPRDDQYVPQCDDLGHFIPLQCHGKSDFCWCVDKDGREVQGTRSQPGTTPACIPTVAPPMVRPTPRPDVTPPSVGTFLLYTQGQQIGYLPLNGTRLQKDAAKTLLSLHGSIIVGIDYDCRERMVYWTDVAGRTISRAGLELGAEPETIVNSGLISPEGLAIDHIRRTMYWTDSVLDKIESALLDGSERKVLFYTDLVNPRAIAVDPIRGNLYWTDWNREAPKIETSSLDGENRRILINTDIGLPNGLTFDPFSKLLCWADAGTKKLECTLPDGTGRRVIQNNLKYPFSIVSYADHFYHTDWRRDGVVSVNKHSGQFTDEYLPEQRSHLYGITAVYPYCPTGRKSpecifications
AAH35608.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
133.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
MEGDRVAGRPVLSSLPVLLLLQLLMLRAAALHPDELFPHGESWGDQLLQEGDDESSAVVKLANPLHFYEARFSNLYVGTNGIISTQDFPRETQYVDYDFPTDFPAIAPFLADIDTSHGRGRVLYRVNGKVSGHLHVGHTPVHFTDVDLHAYIVGNDGRAYTAISHIPQPAAQALLPLTPIGGLFGWLFALEKPGSENGFSLAGAAFTHDMEVTFYPGEETVRITQTAEGLDPENYLSIKTNIQGQVPYVPANFTAHISPYKELYHYSDSTVTSTSSRDYSLTFGAINQTWSYRIHQNITYQVCRHAPRHPSFPTTQQLNVDRVFALYNDEERVLRFAVTNQIGPVKEDSDPTPVNPCYDGSHMCDTTARCHPGTGVDYTCECASGYQGDGRNCVDENECATGFHRCGPNSVCINLPGSYRCECRSAYEFADDRHTCILITPPANPCEDGSHTCAPAGQARCVHHGGSTFSCACLPGYAGDGHQCTDVDECSENRCHPAATCYNTPGSFSCRCQPGYYGDGFQCIPDSTSSLTPCEQQQRHAQAQYAYPGARFHIPQCDEQGNFLPLQCHGSTGFCWCVDPDGHEVPGTQTPPGSTPPHCGPSPEPTQRPPTICERWRENLLEHYGGTPRDDQYVPQCDDLGHFIPLQCHGKSDFCWCVDKDGREVQGTRSQPGTTPACIPTVAPPMVRPTPRPDVTPPSVGTFLLYTQGQQIGYLPLNGTRLQKDAAKTLLSLHGSIIVGIDYDCRERMVYWTDVAGRTISRAGLELGAEPETIVNSGLISPEGLAIDHIRRTMYWTDSVLDKIESALLDGSERKVLFYTDLVNPRAIAVDPIRGNLYWTDWNREAPKIETSSLDGENRRILINTDIGLPNGLTFDPFSKLLCWADAGTKKLECTLPDGTGRRVIQNNLKYPFSIVSYADHFYHTDWRRDGVVSVNKHSGQFTDEYLPEQRSHLYGITAVYPYCPTGRK | |
RUO | |
NID2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
22795 | |
NID2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NID2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |