missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human NGLY1 Full-length ORF (NP_060767.2, 1 a.a. - 654 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00055768-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16121759

  • 6245.00 NOK / 25µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes an enzyme that catalyzes hydrolysis of an N(4)-(acetyl-beta-D-glucosaminyl) asparagine residue to N-acetyl-beta-D-glucosaminylamine and a peptide containing an aspartate residue. The encoded enzyme may play a role in the proteasome-mediated degradation of misfolded glycoproteins. Multiple transcript variants encoding different isoforms have been found for this gene

Sequence: MAAAALGSSSGSASPAVAELCQNTPETFLEASKLLLTYADNILRNPNDEKYRSIRIGNTAFSTRLLPVRGAVECLFEMGFEEGETHLIFPKKASVEQLQKIRDLIAIERSSRLDGSNKSHKVKSSQQPAASTQLPTTPSSNPSGLNQHTRNRQGQSSDPPSASTVAADSAILEVLQSNIQHVLVYENPALQEKALACIPVQELKRKSQEKLSRARKLDKGINISDEDFLLLELLHWFKEEFFHWVNNVLCSKCGGQTRSRDRSLLPSDDELKWGAKEVEDHYCDACQFSNRFPRYNNPEKLLETRCGRCGEWANCFTLCCRAVGFEARYVWDYTDHVWTEVYSPSQQRWLHCDACEDVCDKPLLYEIGWGKKLSYVIAFSKDEVVDVTWRYSCKHEEVIARRTKVKEALLRDTINGLNKQRQLFLSENRRKELLQRIIVELVEFISPKTPKPGELGGRISGSVAWRVARGEMGLQRKETLFIPCENEKISKQLHLCYNIVKDRYVRVSNNNQTISGWENGVWKMESIFRKVETDWHMVYLARKEGSSFAYISWKFECGSVGLKVDSISIRTSSQTFQTGTVEWKLRSDTAQVELTGDNSLHSYADFSGATEVILEAELSRGDGDVAWQHTQLFRQSLNDHEENCLEIIIKFSDL
Specifications

Specifications

NP_060767.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
100.8kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ11005/FLJ12409/PNG1
NGLY1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
55768
NGLY1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAAAALGSSSGSASPAVAELCQNTPETFLEASKLLLTYADNILRNPNDEKYRSIRIGNTAFSTRLLPVRGAVECLFEMGFEEGETHLIFPKKASVEQLQKIRDLIAIERSSRLDGSNKSHKVKSSQQPAASTQLPTTPSSNPSGLNQHTRNRQGQSSDPPSASTVAADSAILEVLQSNIQHVLVYENPALQEKALACIPVQELKRKSQEKLSRARKLDKGINISDEDFLLLELLHWFKEEFFHWVNNVLCSKCGGQTRSRDRSLLPSDDELKWGAKEVEDHYCDACQFSNRFPRYNNPEKLLETRCGRCGEWANCFTLCCRAVGFEARYVWDYTDHVWTEVYSPSQQRWLHCDACEDVCDKPLLYEIGWGKKLSYVIAFSKDEVVDVTWRYSCKHEEVIARRTKVKEALLRDTINGLNKQRQLFLSENRRKELLQRIIVELVEFISPKTPKPGELGGRISGSVAWRVARGEMGLQRKETLFIPCENEKISKQLHLCYNIVKDRYVRVSNNNQTISGWENGVWKMESIFRKVETDWHMVYLARKEGSSFAYISWKFECGSVGLKVDSISIRTSSQTFQTGTVEWKLRSDTAQVELTGDNSLHSYADFSGATEVILEAELSRGDGDVAWQHTQLFRQSLNDHEENCLEIIIKFSDL
RUO
NGLY1
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers