Learn More
Abnova™ Human NFKBIL2 Partial ORF (NP_038460, 1 a.a. - 97 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00004796-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is thought to be a negative regulator of NF-kappa-B mediated transcription. The encoded protein may bind NF-kappa-B complexes and trap them in the cytoplasm, preventing them from entering the nucleus and interacting with the DNA. Phosphorylation of this protein targets it for degradation by the ubiquitination pathway, which frees the NF-kappa-B complexes to enter the nucleus. [provided by RefSeq]
Sequence: MRTRLYLNLGLTFESLQQTALCNDYFRKSIFLAEQNHLYEDLFRARYNLGTIHWRAGQHSQAMRCLEGARECAHTMRKRFMESECCVVIAQVLQDLGSpecifications
NP_038460 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.41kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MRTRLYLNLGLTFESLQQTALCNDYFRKSIFLAEQNHLYEDLFRARYNLGTIHWRAGQHSQAMRCLEGARECAHTMRKRFMESECCVVIAQVLQDLG | |
RUO | |
NFKBIL2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
4796 | |
NFKBIL2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ40087/IKBR | |
NFKBIL2 | |
Recombinant | |
wheat germ expression system |