Learn More
Abnova™ Human NEUROD6 Full-length ORF (AAH35048, 1 a.a. - 337 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00063974-P01.25ug
Additional Details : Weight : 0.00010kg
Description
NEUROD6 is a member of the NEUROD (NEUROD1; MIM 601724) family of basic helix-loop-helix (bHLH) transcription factors (Guo et al., 2002 [PubMed 12357074]).[supplied by OMIM]
Sequence: MLTLPFDESVVMPESQMCRKFSRECEDQKQIKKPESFSKQIVLRGKSIKRAPGEETEKEEEEEDREEEDENGLPRRRGLRKKKTTKLRLERVKFRRQEANARERNRMHGLNDALDNLRKVVPCYSKTQKLSKIETLRLAKNYIWALSEILRIGKRPDLLTFVQNLCKGLSQPTTNLVAGCLQLNARSFMMGQGGEAAHHTRSPYSTFYPPYHSPELTTPPGHGTLDNSKSMKPYNYCSAYESFYESTSPECASPQFEGPLSPPPINYNGIFSLKQEETLDYGKNYNYGMHYCAVPPRGPLGQGAMFRLPTDSHFPYDLHLRSQSLTMQDELNAVFHNSpecifications
AAH35048 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
62.81kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
Atoh2/MATH2/Math-2/NEX1M/bHLHa2 | |
NEUROD6 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
63974 | |
NEUROD6 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MLTLPFDESVVMPESQMCRKFSRECEDQKQIKKPESFSKQIVLRGKSIKRAPGEETEKEEEEEDREEEDENGLPRRRGLRKKKTTKLRLERVKFRRQEANARERNRMHGLNDALDNLRKVVPCYSKTQKLSKIETLRLAKNYIWALSEILRIGKRPDLLTFVQNLCKGLSQPTTNLVAGCLQLNARSFMMGQGGEAAHHTRSPYSTFYPPYHSPELTTPPGHGTLDNSKSMKPYNYCSAYESFYESTSPECASPQFEGPLSPPPINYNGIFSLKQEETLDYGKNYNYGMHYCAVPPRGPLGQGAMFRLPTDSHFPYDLHLRSQSLTMQDELNAVFHN | |
RUO | |
NEUROD6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |