missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human NEGR1 Partial ORF (NP_776169, 228 a.a. - 327 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00257194-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16122647

  • 5540.00 NOK / 25µg
Estimated Shipment: 19-05-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: QEIKSGTVTPGRSGLIRCEGAGVPPPAFEWYKGEKKLFNGQQGIIIQNFSTRSILTVTNVTQEHFGNYTCVAANKLGTTNASLPLNPPSTAQYGITGSAD
Specifications

Specifications

NP_776169
Liquid
257194
NEGR1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DMML2433/IGLON4/KILON/MGC46680/Ntra
NEGR1
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
QEIKSGTVTPGRSGLIRCEGAGVPPPAFEWYKGEKKLFNGQQGIIIQNFSTRSILTVTNVTQEHFGNYTCVAANKLGTTNASLPLNPPSTAQYGITGSAD
RUO
NEGR1
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers