Learn More
Abnova™ Human NEDD9 Partial ORF (NP_006394, 101 a.a. - 200 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand:  Abnova™ H00004739-Q01.10ug
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16128281
  • 4115.00 NOK / 10µg
Estimated Shipment: 01-07-2024
Description
Sequence: PRDTIYQVPPSYQNQGIYQVPTGHGTQEQEVYQVPPSVQRSIGGTSGPHVGKKVITPVRTGHGYVYEYPSRYQKDVYDIPPSHTTQGVYDIPPSSAKGPV
Specifications
Specifications
NP_006394
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
PRDTIYQVPPSYQNQGIYQVPTGHGTQEQEVYQVPPSVQRSIGGTSGPHVGKKVITPVRTGHGYVYEYPSRYQKDVYDIPPSHTTQGVYDIPPSSAKGPV
RUO
NEDD9
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
4739
NEDD9 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CAS-L/CAS2/CASL/CASS2/HEF1/dJ49G10.2/dJ761I2.1
NEDD9
Recombinant
wheat germ expression system