missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ND1 Partial ORF (YP_003024026, 21 a.a. - 71 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00004535-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
Sequence: MLTERKILGYIQLRKGPNVVGPYGLLQPFADAIKLFTKEPLKPATSTITLYSpecifications
YP_003024026 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
31.35kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MLTERKILGYIQLRKGPNVVGPYGLLQPFADAIKLFTKEPLKPATSTITLY | |
RUO | |
ND1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
4535 | |
ND1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MTND1 | |
ND1 | |
Recombinant | |
wheat germ expression system |