missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human NCAPH2 Full-length ORF (NP_689512.1, 1 a.a. - 555 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00029781-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16129102

  • 4115.00 NOK / 10µg
Estimated Shipment: 26-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Condensin complexes I and II play essential roles in mitotic chromosome assembly and segregation. Both condensins contain 2 invariant structural maintenance of chromosome (SMC) subunits, SMC2 (MIM 605576) and SMC4 (MIM 605575), but they contain different sets of non-SMC subunits. NCAPH2 is 1 of 3 non-SMC subunits that define condensin II (Ono et al., 2003 [PubMed 14532007]).[supplied by OMIM]

Sequence: MNFIEAALLIQGSACVYSKKVEYLYSLVYQALDFISGKRRAKQLSSVQEDRANGVASSGVPQEAENEFLSLDDFPDSRTNVDLKNDQTPSEVLIIPLLPMALVAPDEMEKNNNPLYSRQGEVLASRKDFRMNTCVPHPRGAFMLEPEGMSPMEPAGVSPMPGTQKDTGRTEEQPMEVSVCRSPVPALGFSQEPGPSPEGPMPLGGGEDEDAEEAVELPEASAPKAALEPKESRSPQQSAALPRRYMLREREGAPEPASCVKETPDPWQSLDPFDSLESKPFKKGRPYSVPPCVEEALGQKRKRKGAAKLQDFHQWYLAAYADHADSRRLRRKGPSFADMEVLYWTHVKEQLETLRKLQRREVAEQWLRPAEEDHLEDSLEDLGAADDFLEPEEYMEPEGADPREAADLDAVPMSLSYEELVRRNVELFIATSQKFVQETELSQRIRDWEDTVQPLLQEQEQHVPFDIHTYGDQLVSRFPQLNEWCPFAELVAGQPAFEVCRSMLASLQLANDYTVEITQQPGLEMAVDTMSLRLLTHQRAHKRFQTYAAPSMAQP
Specifications

Specifications

NP_689512.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
88.9kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
384D8-2/384D8_6/CAP-H2/MGC15858/MGC18000/MGC2455/MGC4133/MGC5305/MGC8640/hCAP-H2
NCAPH2
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
29781
NCAPH2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MNFIEAALLIQGSACVYSKKVEYLYSLVYQALDFISGKRRAKQLSSVQEDRANGVASSGVPQEAENEFLSLDDFPDSRTNVDLKNDQTPSEVLIIPLLPMALVAPDEMEKNNNPLYSRQGEVLASRKDFRMNTCVPHPRGAFMLEPEGMSPMEPAGVSPMPGTQKDTGRTEEQPMEVSVCRSPVPALGFSQEPGPSPEGPMPLGGGEDEDAEEAVELPEASAPKAALEPKESRSPQQSAALPRRYMLREREGAPEPASCVKETPDPWQSLDPFDSLESKPFKKGRPYSVPPCVEEALGQKRKRKGAAKLQDFHQWYLAAYADHADSRRLRRKGPSFADMEVLYWTHVKEQLETLRKLQRREVAEQWLRPAEEDHLEDSLEDLGAADDFLEPEEYMEPEGADPREAADLDAVPMSLSYEELVRRNVELFIATSQKFVQETELSQRIRDWEDTVQPLLQEQEQHVPFDIHTYGDQLVSRFPQLNEWCPFAELVAGQPAFEVCRSMLASLQLANDYTVEITQQPGLEMAVDTMSLRLLTHQRAHKRFQTYAAPSMAQP
RUO
NCAPH2
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers