Learn More
Abnova™ Human NALP12 Partial ORF (AAH28069, 1 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00091662-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the CATERPILLER family of cytoplasmic proteins. The encoded protein, which contains an N-terminal pyrin domain, a NACHT domain, a NACHT-associated domain, and a C-terminus leucine-rich repeat region, functions as an attenuating factor of inflammation by suppressing inflammatory responses in activated monocytes. Alternatively spliced transcript variants encoding distinct isoforms have been described but the full-length nature of some of these has not been determined. [provided by RefSeq]
Sequence: MLRTAGRDGLCRLSTYLEELEAVELKKFKLYLGTATELGEGKIPWGSMEKAGPLEMAQLLITHFGPEEAWRLALSTFERINRKDLWERGQREDLVRDTPPGGPSSLGNQSSpecifications
AAH28069 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MLRTAGRDGLCRLSTYLEELEAVELKKFKLYLGTATELGEGKIPWGSMEKAGPLEMAQLLITHFGPEEAWRLALSTFERINRKDLWERGQREDLVRDTPPGGPSSLGNQS | |
RUO | |
NLRP12 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
91662 | |
NALP12 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CLR19.3/FCAS2/NALP12/PAN6/PYPAF7/RNO/RNO2 | |
NLRP12 | |
Recombinant | |
wheat germ expression system |