missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human N-Terminal Pro-BNP Antibody, R&D Systems™

Mouse Monoclonal Antibody
2280.00 NOK - 5325.00 NOK
Specifications
| Antigen | BNP |
|---|---|
| Clone | 817130R |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18730207
![]() |
R&D Systems
MAB9769-SP |
25 μg |
2280.00 NOK
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18733967
![]() |
R&D Systems
MAB9769-100 |
100 μg |
5325.00 NOK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BNP Monoclonal specifically detects BNP in Human samples. It is validated for ELISA.Specifications
| BNP | |
| ELISA | |
| Monoclonal | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| BNF, BNP, brain type natriuretic peptide, Gamma-brain natriuretic peptide, natriuretic peptide B, natriuretic peptide precursor B, natriuretic peptides B, natriuretic protein, NPPB | |
| E. coli-derived recombinant human Pro-BNP, His27-His134, Accession No. P16860 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Pro-BNP but does not detect mature human BNP peptide in direct ELISAs. |
| 817130R | |
| ELISA | |
| Unconjugated | |
| Human | |
| P16860 | |
| 4879.0 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title