Learn More
Abnova™ Human MYT1 Partial ORF (NP_004526, 586 a.a. - 685 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00004661-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a member of a family of neural specific, zinc finger-containing DNA-binding proteins. The protein binds to the promoter regions of proteolipid proteins of the central nervous system and plays a role in the developing nervous system. [provided by RefSeq]
Sequence: DYASFDAQVFGKRMLAPKIQTSETSPKAFQCFDYSQDAEAAHMAATAILNLSTRCWEMPENLSTKPQDLPSKSVDIEVDENGTLDLSMHKHRKRENAFPSSpecifications
NP_004526 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
DYASFDAQVFGKRMLAPKIQTSETSPKAFQCFDYSQDAEAAHMAATAILNLSTRCWEMPENLSTKPQDLPSKSVDIEVDENGTLDLSMHKHRKRENAFPS | |
RUO | |
MYT1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
4661 | |
MYT1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C20orf36/MTF1/MYTI/PLPB1 | |
MYT1 | |
Recombinant | |
wheat germ expression system |