Learn More
Abnova™ Human MYOM1 Partial ORF (NP_003794.2, 1586 a.a. - 1684 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00008736-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
The giant protein titin, together with its associated proteins, interconnects the major structure of sarcomeres, the M bands and Z discs. The C-terminal end of the titin string extends into the M line, where it binds tightly to M-band constituents of apparent molecular masses of 190 kD (myomesin 1) and 165 kD (myomesin 2). This protein, myomesin 1, like myomesin 2, titin, and other myofibrillar proteins contains structural modules with strong homology to either fibronectin type III (motif I) or immunoglobulin C2 (motif II) domains. Myomesin 1 and myomesin 2 each have a unique N-terminal region followed by 12 modules of motif I or motif II, in the arrangement II-II-I-I-I-I-I-II-II-II-II-II. The two proteins share 50% sequence identity in this repeat-containing region. The head structure formed by these 2 proteins on one end of the titin string extends into the center of the M band. The integrating structure of the sarcomere arises from muscle-specific members of the superfamily of immunoglobulin-like proteins. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]
Sequence: EGKALNLTCNVWGDPPPEVSWLKNEKALASDDHCNLKFEAGRTAYFTINGVSTADSGKYGLVVKNKYGSETSDFTVSVFIPEEEARMAALESLKGGKKASpecifications
NP_003794.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
EGKALNLTCNVWGDPPPEVSWLKNEKALASDDHCNLKFEAGRTAYFTINGVSTADSGKYGLVVKNKYGSETSDFTVSVFIPEEEARMAALESLKGGKKA | |
RUO | |
MYOM1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
8736 | |
MYOM1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC134946/MGC134947/SKELEMIN | |
MYOM1 | |
Recombinant | |
wheat germ expression system |