missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MUTED Full-length ORF (NP_958437.1, 1 a.a. - 187 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00063915-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16183293

  • 4115.00 NOK / 10µg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a component of BLOC-1 (biogenesis of lysosome-related organelles complex 1). Components of this complex are involved in the biogenesis of organelles such as melanosomes and platelet-dense granules. A mouse model for Hermansky-Pudlak Syndrome is mutated in the murine version of this gene. Some transcripts of the downstream gene TXNDC5 overlap this gene, but they do not contain an open reading frame for this gene. [provided by RefSeq]

Sequence: MSGGGTETPVGCEAAPGGGSKKRDSLGTAGSAHLIIKDLGEIHSRLLDHRPVIQGETRYFVKEFEEKRGLREMRVLENLKNMIHETNEHTLPKCRDTMRDSLSQVLQRLQAANDSVCRLQQREQERKKIHSDHLVASEKQHMLQWDNFMKEQPNKRAEVDEEHRKAMERLKEQYAEMEKDLAKFSTF
Specifications

Specifications

NP_958437.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
48kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp686E2287/MU
MUTED
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
63915
MUTED (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSGGGTETPVGCEAAPGGGSKKRDSLGTAGSAHLIIKDLGEIHSRLLDHRPVIQGETRYFVKEFEEKRGLREMRVLENLKNMIHETNEHTLPKCRDTMRDSLSQVLQRLQAANDSVCRLQQREQERKKIHSDHLVASEKQHMLQWDNFMKEQPNKRAEVDEEHRKAMERLKEQYAEMEKDLAKFSTF
RUO
MUTED
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers