missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MPHOSPH1 Partial ORF (NP_057279, 1671 a.a. - 1780 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009585-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Sequence: RSQASIIGVNLATKKKEGTLQKFGDFLQHSPSILQSKAKKIIETMSSSKLSNVEASKENVSQPKRAKRKLYTSEISSPIDISGQVILMDQKMKESDHQIIKRRLRTKTAKSpecifications
NP_057279 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RSQASIIGVNLATKKKEGTLQKFGDFLQHSPSILQSKAKKIIETMSSSKLSNVEASKENVSQPKRAKRKLYTSEISSPIDISGQVILMDQKMKESDHQIIKRRLRTKTAK | |
RUO | |
KIF20B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
9585 | |
MPHOSPH1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp434B0435/DKFZp434P0810/KRMP1/MPHOSPH1/MPP-1/MPP1 | |
KIF20B | |
Recombinant | |
wheat germ expression system |