missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse SUMO3 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF2959
This item is not returnable.
View return policy
Description
SUMO3 Polyclonal specifically detects SUMO3 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| SUMO3 | |
| Polyclonal | |
| Unconjugated | |
| P55854 | |
| SUMO3 | |
| E. coli-derived recombinant human SUMO3 Met1-Phe103 Accession # P55854 | |
| 100 μg | |
| Primary | |
| Detects endogenous human and mouse proteins modified with SUMO3 in Western blots. In Western blots, less than 10% cross-reactivity with recombinant human SUMO2 and SUMO4 is observed, and less than 1% cross-reactivity with SUMO1 is observed. | |
| Human, Mouse | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| SMT3 (suppressor of mif two 3, yeast) homolog 1, SMT3 homolog 1, SMT3 suppressor of mif two 3 homolog 1, SMT3 suppressor of mif two 3 homolog 3 (S. cerevisiae), SMT3 suppressor of mif two 3 homolog 3 (yeast), SMT3A, SMT3B, SMT3H1, SMT3H1small ubiquitin-related modifier 3, SUMO-2, SUMO-3, ubiquitin-like protein SMT3A, Ubiquitin-like protein SMT3B | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 6612 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction