Learn More
Human/Mouse/Rat SOX1 Antibody, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody has been used in 42 publications
Brand:  R&D Systems AF3369
Product Code. 18088981
  • 6030.00 NOK / 100µg
Description
SOX1 Polyclonal specifically detects SOX1 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SOX1
Polyclonal
Unconjugated
NP_005977
SOX1
E. coli-derived recombinant human SOX1 Asn242-Gly379 (Leu276Ile) Accession # NP_005977
100 μg
Primary
Detects human SOX1 in direct ELISAs and Western blots. In direct ELISAs, less than 15% cross-reactivity with recombinant human (rh) SOX2 is observed.
Human, Mouse, Rat
IgG
Western Blot, Immunocytochemistry
LYOPH
Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunocytochemistry 5-15 ug/mL
SRY (sex determining region Y)-box 1, SRY-related HMG-box gene 1, transcription factor SOX-1
Goat
Affinity Purified
RUO
6656
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.