Learn More
Human/Mouse/Rat p53 Agarose-conjugated Antibody, R&D Systems™
Product Code. 18182631
Change view
Click to view available options
Quantity:
250 μL
Unit Size:
250µL
Product Code. Quantity unitSize
18182631 250 μL 250µL
1 options
Product Code. 18182631
Brand: R&D Systems GAF1355
Goat Polyclonal Antibody
p53 Polyclonal specifically detects p53 in Human, Mouse, Rat samples. It is validated for Immunoprecipitation.
TRUSTED_SUSTAINABILITY
Specifications
Antigen p53
Applications Immunoprecipitation
Classification Polyclonal
Conjugate Agarose
Dilution Immunoprecipitation
Gene Alias Antigen NY-CO-13, BCC7, FLJ92943, LFS1, LFS1TRP53, p53, p53 tumor suppressor, P53cellular tumor antigen p53, Phosphoprotein p53, TP53, transformation-related protein 53, TRP53, tumor protein p53, Tumor suppressor p53
Gene Symbols TP53
Host Species Goat
Purification Method Affinity Purified
Quantity 250 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7157
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human, Mouse, Rat
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG