missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human/Mouse/Rat Irisin, Carrier-Free Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ 8880-IR-025/CF
This item is not returnable.
View return policy
Description
The Recombinant Human/Mouse/Rat Irisin Protein is derived from CHO. The Recombinant Human/Mouse/Rat Irisin Protein has been validated for the following applications: Bioactivity.Specifications
| Q8NAU1 | |
| Observed M.W.: 15-37kDa, reducing conditions, Theoretical M.W.: 13kDa | |
| Chinese Hamster Ovary cell line,CHO-derived Irisin/FNDC5 protein Asp32-Glu143,with a C-terminal 6-His tag | |
| <0.10 EU per 1μg of the protein by the LAL method. | |
| Irisin/FNDC5 | |
| Measured by its ability to induce p38 MAPK activation in 3T3-L1 mouse embryonic fibroblast adipose-like cells. 1μg/mL of Recombinant Human Irisin can effectively induce p38 MAPK activation. | |
| Yes |
| 252995 | |
| 25μg | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
| fibronectin type III domain containing 5, fibronectin type III domain-containing protein 5, FNDC5, FRCP2, FRCP2Fibronectin type III repeat-containing protein 2 | |
| FNDC5 | |
| Unconjugated |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?