missing translation for 'onlineSavingsMsg'
Learn More

Human/Mouse Lysine (K)-specific Demethylase 4A/KDM4A Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18756227
Change view
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
Product Code. Quantity unitSize
18756227 25 μg 25µg
1 options
This item is not returnable. View return policy

Product Code. 18756227

Brand: R&D Systems AF6434SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Lysine (K)-specific Demethylase 4A/KDM4A/JMJD2A Polyclonal specifically detects Lysine (K)-specific Demethylase 4A/KDM4A/JMJD2A in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lysine (K)-specific Demethylase 4A/KDM4A/JMJD2A
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. AAH02558
Gene Alias EC 1.14.11, EC 1.14.11.-, JHDM3A, JHDM3AJMJD2, JmjC domain-containing histone demethylation protein 3A, JMJD2, JMJD2A, JMJD2Alysine-specific demethylase 4A, jumonji domain containing 2, jumonji domain containing 2A, Jumonji domain-containing protein 2A, KDM4A, KIAA0677jumonji C domain-containing histone demethylase 3A, lysine (K)-specific demethylase 4A, Lysine (K)specific Demethylase 4A
Gene Symbols KDM4A
Host Species Sheep
Immunogen E. coli-derived recombinant human KDM4A Lys386-Ser514 Accession # AAH02558
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9682
Test Specificity Detects human KDM4A in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human KDM4C is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.