missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Brachyury Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 94 publications
Brand: R&D Systems AF2085
This item is not returnable.
View return policy
Description
Brachyury Polyclonal specifically detects Brachyury in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Chromatin Immunoprecipitation (ChIP), Immunocytochemistry.
Specifications
| Brachyury | |
| Polyclonal | |
| Unconjugated | |
| O15178 | |
| T | |
| E. coli-derived recombinant human Brachyury Ser2-Glu202 Accession # O15178 | |
| 100 μg | |
| Primary | |
| Detects human Brachyury in direct ELISAs and Western blots. | |
| Human, Mouse | |
| IgG |
| ChIP Assay, Immunocytochemistry, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1-1 ug/mL, Immunohistochemistry 5-15 ug/mL, Chromatin Immunoprecipitation (ChIP) 5 ug/10^6 cells, Immunocytochemistry 5-15 ug/mL | |
| brachyury protein, MGC104817, Protein T, SAVA, T brachyury (mouse) homolog, T brachyury homolog, T Brachyury Transcription Factor, T, brachyury homolog (mouse), T-Box Transcription Factor T, TBXT, TF, TFT | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 6862 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction