missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MMP23B Partial ORF (NP_008914, 241 a.a. - 340 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00008510-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16128695

  • 5540.00 NOK / 25µg
Estimated Shipment: 22-05-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene (MMP23B) encodes a member of the matrix metalloproteinase (MMP) family, and it is part of a duplicated region of chromosome 1p36.3. Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. This gene belongs to the more telomeric copy of the duplicated region. [provided by RefSeq]

Sequence: LSQDELWGLHRLYGCLDRLFVCASWARRGFCDARRRLMKRLCPSSCDFCYEFPFPTVATTPPPPRTKTRLVPEGRNVTFRCGQKILHKKGKVYWYKDQEP
Specifications

Specifications

NP_008914
Liquid
8510
MMP23B (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MIFR/MIFR-1/MMP22
MMP23B
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
LSQDELWGLHRLYGCLDRLFVCASWARRGFCDARRRLMKRLCPSSCDFCYEFPFPTVATTPPPPRTKTRLVPEGRNVTFRCGQKILHKKGKVYWYKDQEP
RUO
MMP23B
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers