Learn More
Abnova™ Human MINA Full-length ORF (AAH14928.1, 1 a.a. - 465 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00084864-P01.25ug
Additional Details : Weight : 0.00010kg
Description
MINA is a c-Myc (MYC; MIM 190080) target gene that may play a role in cell proliferation or regulation of cell growth. (Tsuneoka et al., 2002 [PubMed 12091391]; Zhang et al., 2005 [PubMed 15897898]).[supplied by OMIM]
Sequence: MPKKAKPTGSGKEEGPAPCKQMKLEAAGGPSALNFDSPSSLFESLISPIKTETFFKEFWEQKPLLIQRDDPALATYYGSLFKLTDLKSLCSRGMYYGRDVNVCRCVNGKKKVLNKDGKAHFLQLRKDFDQKRATIQFHQPQRFKDELWRIQEKLECYFGSLVGSNVYITPAGSQGLPPHYDDVEVFILQLEGEKHWRLYHPTVPLAREYSVEAEERIGRPVHEFMLKPGDLLYFPRGTIHQADTPAGLAHSTHVTISTYQNNSWGDFLLDTISGLVFDTAKEDVELRTGIPRQLLLQVESTTVATRRLSGFLRTLADRLEGTKELLSSDMKKDFIMHRLPPYSAGDGAELSTPGGKLPRLDSVVRLQFKDHIVLTVLPDQDQSDETQEKMVYIYHSLKNSRETHMMGNEEETEFHGLRFPLSHLDALKQIWNSPAISVKDLKLTTDEEKESLVLSLWTECLIQVVSpecifications
AAH14928.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
79.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp762O1912/FLJ14393/MDIG/MINA53/NO52 | |
MINA | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
84864 | |
MINA (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MPKKAKPTGSGKEEGPAPCKQMKLEAAGGPSALNFDSPSSLFESLISPIKTETFFKEFWEQKPLLIQRDDPALATYYGSLFKLTDLKSLCSRGMYYGRDVNVCRCVNGKKKVLNKDGKAHFLQLRKDFDQKRATIQFHQPQRFKDELWRIQEKLECYFGSLVGSNVYITPAGSQGLPPHYDDVEVFILQLEGEKHWRLYHPTVPLAREYSVEAEERIGRPVHEFMLKPGDLLYFPRGTIHQADTPAGLAHSTHVTISTYQNNSWGDFLLDTISGLVFDTAKEDVELRTGIPRQLLLQVESTTVATRRLSGFLRTLADRLEGTKELLSSDMKKDFIMHRLPPYSAGDGAELSTPGGKLPRLDSVVRLQFKDHIVLTVLPDQDQSDETQEKMVYIYHSLKNSRETHMMGNEEETEFHGLRFPLSHLDALKQIWNSPAISVKDLKLTTDEEKESLVLSLWTECLIQVV | |
RUO | |
MINA | |
Wheat Germ (in vitro) | |
GST | |
Liquid |