missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MIB1 Partial ORF (NP_065825, 909 a.a. - 1006 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00057534-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16142763

  • 6245.00 NOK / 25µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: PFIMCCGGKSSEDATDDISSGNIPVLQKDKDNTNVNADVQKLQQQLQDIKEQTMCPVCLDRLKNMIFLCGHGTCQLCGDRMSECPICRKAIERRILLY
Specifications

Specifications

NP_065825
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.52kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
PFIMCCGGKSSEDATDDISSGNIPVLQKDKDNTNVNADVQKLQQQLQDIKEQTMCPVCLDRLKNMIFLCGHGTCQLCGDRMSECPICRKAIERRILLY
RUO
MIB1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
57534
MIB1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DIP-1/DKFZp686I0769/DKFZp761M1710/FLJ90676/MGC129659/MGC129660/MIB/ZZANK2/ZZZ6
MIB1
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers