missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MIA2 Partial ORF (NP_473365.3, 448 a.a. - 553 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00117153-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16131557

  • 4115.00 NOK / 10µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: TDESDTIPYLKKFLYNFDNPWNFQNIPKETELPFPKQILDQNNVIENEETGEFSIDNYPTDNTKVMIFKSSYSLSDMVSNIELPTRIHEEVYFEPSSSKDSDENSK
Specifications

Specifications

NP_473365.3
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.4kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
TDESDTIPYLKKFLYNFDNPWNFQNIPKETELPFPKQILDQNNVIENEETGEFSIDNYPTDNTKVMIFKSSYSLSDMVSNIELPTRIHEEVYFEPSSSKDSDENSK
RUO
MIA2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
117153
MIA2 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ22404
MIA2
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers