Learn More
Abnova™ Human MGAT4B Full-length ORF (NP_463459.1, 1 a.a. - 563 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00011282-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a key glycosyltransferase that regulates the formation of tri- and multiantennary branching structures in the Golgi apparatus. The encoded protein, in addition to the related isoenzyme A, catalyzes the transfer of N-acetylglucosamine (GlcNAc) from UDP-GlcNAc in a beta-1,4 linkage to the Man-alpha-1,3-Man-beta-1,4-GlcNAc arm of R-Man-alpha-1,6(GlcNAc-beta-1,2-Man-alpha-1,3)Man-beta-1,4-GlcNAc-beta-1,4-GlcNAc-beta-1-Asn. The encoded protein may play a role in regulating the availability of serum glycoproteins, oncogenesis, and differentiation. [provided by RefSeq]
Sequence: MSRVAGTRTDVNELLQRWTPRCVRWHTGGARRVALDRPLVTACLPPAGDVVDVYQREFLALRDRLHAAEQESLKRSKELNLVLDEIKRAVSERQALRDGDGNRTWGRLTEDPRLKPWNGSHRHVLHLPTVFHHLPHLLAKESSLQPAVRVGQGRTGVSVVMGIPSVRREVHSYLTDTLHSLISELSPQEKEDSVIVVLIAETDSQYTSAVTENIKALFPTEIHSGLLEVISPSPHFYPDFSRLRESFGDPKERVRWRTKQNLDYCFLMMYAQSKGIYYVQLEDDIVAKPNYLSTMKNFALQQPSEDWMILEFSQLGFIGKMFKSLDLSLIVEFILMFYRDKPIDWLLDHILWVKVCNPEKDAKHCDRQKANLRIRFKPSLFQHVGTHSSLAGKIQKLKDKDFGKQALRKEHVNPPAEVSTSLKTYQHFTLEKAYLREDFFWAFTPAAGDFIRFRFFQPLRLERFFFRSGNIEHPEDKLFNTSVEVLPFDNPQSDKEALQEGRTATLRYPRSPDGYLQIGSFYKGVAEGEVDPAFGPLEALRLSIQTDSPVWVILSEIFLKKADSpecifications
NP_463459.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
91.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GNT-IV/GNT-IVB | |
MGAT4B | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
11282 | |
MGAT4B (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSRVAGTRTDVNELLQRWTPRCVRWHTGGARRVALDRPLVTACLPPAGDVVDVYQREFLALRDRLHAAEQESLKRSKELNLVLDEIKRAVSERQALRDGDGNRTWGRLTEDPRLKPWNGSHRHVLHLPTVFHHLPHLLAKESSLQPAVRVGQGRTGVSVVMGIPSVRREVHSYLTDTLHSLISELSPQEKEDSVIVVLIAETDSQYTSAVTENIKALFPTEIHSGLLEVISPSPHFYPDFSRLRESFGDPKERVRWRTKQNLDYCFLMMYAQSKGIYYVQLEDDIVAKPNYLSTMKNFALQQPSEDWMILEFSQLGFIGKMFKSLDLSLIVEFILMFYRDKPIDWLLDHILWVKVCNPEKDAKHCDRQKANLRIRFKPSLFQHVGTHSSLAGKIQKLKDKDFGKQALRKEHVNPPAEVSTSLKTYQHFTLEKAYLREDFFWAFTPAAGDFIRFRFFQPLRLERFFFRSGNIEHPEDKLFNTSVEVLPFDNPQSDKEALQEGRTATLRYPRSPDGYLQIGSFYKGVAEGEVDPAFGPLEALRLSIQTDSPVWVILSEIFLKKAD | |
RUO | |
MGAT4B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |