missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MFRP Partial ORF (NP_113621, 480 a.a. - 579 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00083552-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16140487

  • 6245.00 NOK / 25µg
Estimated Shipment: 19-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a member of the frizzled-related proteins. The encoded protein may play a role in eye development, as mutations in this gene have been associated with nanophthalmos, posterior microphthalmia, retinitis pigmentosa, foveoschisis, and optic disc drusen. The protein is encoded by a bicistronic mRNA, which also encodes C1q and tumor necrosis factor related protein 5

Sequence: NTTAFPNIWVGMITQEEVVEVLSGYKSLTSLPCYQHFRRLLCGLLVPRCTPLGSVLPPCRSVCQEAEHQCQSGLALLGTPWPFNCNRLPEAADLEACAQP
Specifications

Specifications

NP_113621
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
NTTAFPNIWVGMITQEEVVEVLSGYKSLTSLPCYQHFRRLLCGLLVPRCTPLGSVLPPCRSVCQEAEHQCQSGLALLGTPWPFNCNRLPEAADLEACAQP
RUO
MFRP
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
83552
MFRP (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C1QTNF5/FLJ30570/NNO2/rd6
MFRP
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers