missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human METTL2B Full-length ORF (AAI07587.1, 1 a.a. - 313 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00055798-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16141823

  • 6270.00 NOK / 25µg
Estimated Shipment: 01-12-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene is a member of a family of methyltransferases that share homology with, but are distinct from, the UbiE family of methyltransferases. Alternatively spliced variants which encode different protein isoforms have been described; however, not all variants have been fully characterized. [provided by RefSeq]

Sequence: MPVDYEINAHKYWNDFYKIHENGFFKDRHWLFTEFPELAPSQNQNHLKDWFLENKSEVCECRNNEDGPGLIMEEQHKCSSKSLEHKTQTPPVEENVTQKISDLEICADEFPGSSATYRILEVGCGVGNTVFPILQTNNDPGLFVYCCDFSSTAIELVQTNSEYDPSRCFAFVHDLCDEEKSYPVPKGSLDIIILIFVLSAIVPDKMQKAINRLSRLLKPGGMVLLRDYGRYDMAQLRFKKGQCLSGNFYVRGDGTRVYFFTQEELDTLFTTAGLEKVQNLVDRRLQVNRGKQLTMYRVWIQCKYCKPLLSSTS
Specifications

Specifications

AAI07587.1
Liquid
55798
METTL2B (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MPVDYEINAHKYWNDFYKIHENGFFKDRHWLFTEFPELAPSQNQNHLKDWFLENKSEVCECRNNEDGPGLIMEEQHKCSSKSLEHKTQTPPVEENVTQKISDLEICADEFPGSSATYRILEVGCGVGNTVFPILQTNNDPGLFVYCCDFSSTAIELVQTNSEYDPSRCFAFVHDLCDEEKSYPVPKGSLDIIILIFVLSAIVPDKMQKAINRLSRLLKPGGMVLLRDYGRYDMAQLRFKKGQCLSGNFYVRGDGTRVYFFTQEELDTLFTTAGLEKVQNLVDRRLQVNRGKQLTMYRVWIQCKYCKPLLSSTS
RUO
METTL2B
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
62.5kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ11350/FLJ12760/METL/METTL2/METTL2A/PSENIP1
METTL2B
Yes
wheat germ expression system
Videos
SDS
Documents
Certificates
Special Offers

Special Offers