Learn More
Abnova™ Human MED8 Partial ORF (NP_443109, 1 a.a. - 59 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00112950-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein that is one of more than 20 subunits of the mediator complex, first identified in S. cerevisiae, that is required for activation of transcription. The product of this gene also interacts with elongins B and C, and CUL2 and RBX1, to reconstitute a ubiquitin ligase. Two alternative transcripts encoding different isoforms have been described. [provided by RefSeq]
Sequence: MRQTEGRVPVFSHEVVPDHLRTKPDPEVEEQEKQLTTDAARIGADAAQKQIQSLNKMCSSpecifications
NP_443109 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
32.23kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MRQTEGRVPVFSHEVVPDHLRTKPDPEVEEQEKQLTTDAARIGADAAQKQIQSLNKMCS | |
RUO | |
MED8 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
112950 | |
MED8 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ARC32/MGC17544/MGC19641 | |
MED8 | |
Recombinant | |
wheat germ expression system |