missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MDFI Partial ORF (NP_005577, 33 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00004188-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16191085

  • 4115.00 NOK / 10µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This protein is a transcription factor that negatively regulates other myogenic family proteins. Studies of the mouse homolog, I-mf, show that it interferes with myogenic factor function by masking nuclear localization signals and preventing DNA binding. Knockout mouse studies show defects in the formation of vertebrae and ribs that also involve cartilage formation in these structures. [provided by RefSeq]

Sequence: PGLEVVTGSTHPAEAAPEEGSLEEAATPMPQGNGPGIPQGLDSTDLDVPTEAVTCQPQGNPLGCTPLLPNDSGHPSEL
Specifications

Specifications

NP_005577
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
34.32kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
PGLEVVTGSTHPAEAAPEEGSLEEAATPMPQGNGPGIPQGLDSTDLDVPTEAVTCQPQGNPLGCTPLLPNDSGHPSEL
RUO
MDFI
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
4188
MDFI (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
I-MF/I-mfa
MDFI
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers