missing translation for 'onlineSavingsMsg'
Learn More

Human MBL Antibody, R&D Systems™
GREENER_CHOICE

Product Code. p-7064987
Change view
Click to view available options
Quantity:
25 μg
500 μg
Unit Size:
25µg
500µg
Product Code. Quantity unitSize
18743808 25 μg 25µg
18130701 500 μg 500µg
2 options
This item is not returnable. View return policy

Product Code. 18743808

Brand: R&D Systems MAB23071SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MBL Monoclonal specifically detects MBL in Human samples. It is validated for Blockade of Receptor-ligand Interaction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MBL
Applications Blockade of Receptor-ligand Interaction
Classification Monoclonal
Clone 285623
Conjugate Unconjugated
Dilution Blockade of Receptor-ligand Interaction
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. AAH96182
Gene Alias COLEC1, COLEC1collectin-1, Collectin-1, HSMBPC, Mannan-binding protein, Mannose-binding lectin, mannose-binding lectin (protein C) 2, soluble, mannose-binding lectin (protein C) 2, soluble (opsonic defect), mannose-binding protein C, MBL2, MBL2D, MBLmannan-binding lectin, MBP, MBP1, MBP1mannose-binding lectin 2, soluble (opsonic defect), MBP-C, MGC116832, MGC116833
Gene Symbols MBL2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human MBL Glu21-Ile248 Accession # AAH96182
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4153
Test Specificity Detects human MBL in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse MBL-2 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.