missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MASP2 Full-length ORF (NP_006601.2, 1 a.a. - 686 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00010747-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16125652

  • 4115.00 NOK / 10µg
Estimated Shipment: 26-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Ra-reactive factor (RARF) is a complement-dependent bactericidal factor that binds to the Ra and R2 polysaccharides expressed by certain enterobacteria. Alternate splicing of this gene results in two transcript variants encoding two RARF components that are involved in the mannan-binding lectin pathway of complement activation. The longer isoform is cleaved into two chains which form a heterodimer linked by a disulfide bond. The encoded proteins are members of the trypsin family of peptidases. [provided by RefSeq]

Sequence: MRLLTLLGLLCGSVATPLGPKWPEPVFGRLASPGFPGEYANDQERRWTLTAPPGYRLRLYFTHFDLELSHLCEYDFVKLSSGAKVLATLCGQESTDTERAPGKDTFYSLGSSLDITFRSDYSNEKPFTGFEAFYAAEDIDECQVAPGEAPTCDHHCHNHLGGFYCSCRAGYVLHRNKRTCSALCSGQVFTQRSGELSSPEYPRPYPKLSSCTYSISLEEGFSVILDFVESFDVETHPETLCPYDFLKIQTDREEHGPFCGKTLPHRIETKSNTVTITFVTDESGDHTGWKIHYTSTAQPCPYPMAPPNGHVSPVQAKYILKDSFSIFCETGYELLQGHLPLKSFTAVCQKDGSWDRPMPACSIVDCGPPDDLPSGRVEYITGPGVTTYKAVIQYSCEETFYTMKVNDGKYVCEADGFWTSSKGEKSLPVCEPVCGLSARTTGGRIYGGQKAKPGDFPWQVLILGGTTAAGALLYDNWVLTAAHAVYEQKHDASALDIRMGTLKRLSPHYTQAWSEAVFIHEGYTHDAGFDNDIALIKLNNKVVINSNITPICLPRKEAESFMRTDDIGTASGWGLTQRGFLARNLMYVDIPIVDHQKCTAAYEKPPYPRGSVTANMLCAGLESGGKDSCRGDSGGALVFLDSETERWFVGGIVSWGSMNCGEAGQYGVYTKVINYIPWIENIISDF
Specifications

Specifications

NP_006601.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
102.1kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MAP19/MASP-2/sMAP
MASP2
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
10747
MASP2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MRLLTLLGLLCGSVATPLGPKWPEPVFGRLASPGFPGEYANDQERRWTLTAPPGYRLRLYFTHFDLELSHLCEYDFVKLSSGAKVLATLCGQESTDTERAPGKDTFYSLGSSLDITFRSDYSNEKPFTGFEAFYAAEDIDECQVAPGEAPTCDHHCHNHLGGFYCSCRAGYVLHRNKRTCSALCSGQVFTQRSGELSSPEYPRPYPKLSSCTYSISLEEGFSVILDFVESFDVETHPETLCPYDFLKIQTDREEHGPFCGKTLPHRIETKSNTVTITFVTDESGDHTGWKIHYTSTAQPCPYPMAPPNGHVSPVQAKYILKDSFSIFCETGYELLQGHLPLKSFTAVCQKDGSWDRPMPACSIVDCGPPDDLPSGRVEYITGPGVTTYKAVIQYSCEETFYTMKVNDGKYVCEADGFWTSSKGEKSLPVCEPVCGLSARTTGGRIYGGQKAKPGDFPWQVLILGGTTAAGALLYDNWVLTAAHAVYEQKHDASALDIRMGTLKRLSPHYTQAWSEAVFIHEGYTHDAGFDNDIALIKLNNKVVINSNITPICLPRKEAESFMRTDDIGTASGWGLTQRGFLARNLMYVDIPIVDHQKCTAAYEKPPYPRGSVTANMLCAGLESGGKDSCRGDSGGALVFLDSETERWFVGGIVSWGSMNCGEAGQYGVYTKVINYIPWIENIISDF
RUO
MASP2
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers