missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MARK3 Partial ORF (NP_002367.4, 402 a.a. - 500 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00004140-Q02.10ug
Additional Details : Weight : 0.02000kg
Description
Sequence: HHKVQRSVFSSQKQRRYSDHAGPAIPSVVAYPKRSQTSTADSDLKEDGISSRKSSGSAVGGKGIAPASPMLGNASNPNKADIPERKKSSTVPSSNTASGSpecifications
NP_002367.4 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
HHKVQRSVFSSQKQRRYSDHAGPAIPSVVAYPKRSQTSTADSDLKEDGISSRKSSGSAVGGKGIAPASPMLGNASNPNKADIPERKKSSTVPSSNTASG | |
RUO | |
MARK3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
4140 | |
MARK3 (Human) Recombinant Protein (Q02) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CTAK1/KP78/PAR1A | |
MARK3 | |
Recombinant | |
wheat germ expression system |