Learn More
Abnova™ Human MAP3K10 Partial ORF (NP_002437, 855 a.a. - 954 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00004294-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a member of the serine/threonine kinase family. This kinase has been shown to activate MAPK8/JNK and MKK4/SEK1, and this kinase itself can be phoshorylated, and thus activated by JNK kinases. This kinase functions preferentially on the JNK signaling pathway, and is reported to be involved in nerve growth factor (NGF) induced neuronal apoptosis. [provided by RefSeq]
Sequence: FPRLPDPQALFPARRRPPEFPGRPTTLTFAPRPRPAASRPRLDPWKLVSFGRTLTISPPSRPDTPESPGPPSVQPTLLDMDMEGQNQDSTVPLCGAHGSHSpecifications
NP_002437 | |
Liquid | |
4294 | |
MAP3K10 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MLK2/MST | |
MAP3K10 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
FPRLPDPQALFPARRRPPEFPGRPTTLTFAPRPRPAASRPRLDPWKLVSFGRTLTISPPSRPDTPESPGPPSVQPTLLDMDMEGQNQDSTVPLCGAHGSH | |
RUO | |
MAP3K10 | |
Wheat Germ (in vitro) | |
GST |