Learn More
Abnova™ Human MAD1L1 Partial ORF (AAH09964, 619 a.a. - 718 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00008379-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
MAD1L1 is a component of the mitotic spindle-assembly checkpoint that prevents the onset of anaphase until all chromosome are properly aligned at the metaphase plate. MAD1L1 functions as a homodimer and interacts with MAD2L1. MAD1L1 may play a role in cell cycle control and tumor suppression. Three transcript variants encoding the same protein have been found for this gene. [provided by RefSeq]
Sequence: KEVFQTKIQEFRKACYTLTGYQIDITTENQYRLTSLYAEHPGDCLIFKATSPSGSKMQLLETEFSHTVGELIEVHLRRQDSIPAFLSSLTLELFSRQTVASpecifications
AAH09964 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
KEVFQTKIQEFRKACYTLTGYQIDITTENQYRLTSLYAEHPGDCLIFKATSPSGSKMQLLETEFSHTVGELIEVHLRRQDSIPAFLSSLTLELFSRQTVA | |
RUO | |
MAD1L1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
8379 | |
MAD1L1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HsMAD1/MAD1/PIG9/TP53I9/TXBP181 | |
MAD1L1 | |
Recombinant | |
wheat germ expression system |