missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human LRRC4C Partial ORF (NP_065980.1, 541 a.a. - 640 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00057689-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16189226

  • 6245.00 NOK / 25µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

NGL1 is a specific binding partner for netrin G1 (NTNG1; MIM 608818), which is a member of the netrin family of axon guidance molecules (Lin et al., 2003 [PubMed 14595443]).[supplied by OMIM]

Sequence: AVMLVIFYKMRKQHHRQNHHAPTRTVEIINVDDEITGDTPMESHLPMPAIEHEHLNHYNSYKSPFNHTTTVNTINSIHSSVHEPLLIRMNSKDNVQETQI
Specifications

Specifications

NP_065980.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
AVMLVIFYKMRKQHHRQNHHAPTRTVEIINVDDEITGDTPMESHLPMPAIEHEHLNHYNSYKSPFNHTTTVNTINSIHSSVHEPLLIRMNSKDNVQETQI
RUO
LRRC4C
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
57689
LRRC4C (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
KIAA1580/NGL-1/NGL1
LRRC4C
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers