missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human LRCH4 Partial ORF (NP_002310.2, 274 a.a. - 379 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00004034-Q01.10ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16094295

  • 4115.00 NOK / 10µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a protein that contains leucine-rich repeats (LRR) at its amino terminus and that is known to be involved in ligand binding. The carboxyl terminus may act as a membrane anchor. Identified structural elements suggest that the encoded protein resembles a receptor. [provided by RefSeq]

Sequence: PSRPPSFSPCPAEDLFPGHRYDGGLDSGFHSVDSGSKRWSGNESTDEFSELSFRISELAREPRGPRERKEDGSADGDPVQIDFIDSHVPGEDEERGTVEEQRPPEL
Specifications

Specifications

NP_002310.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.29kDa
12.5% SDS-PAGE Stained with Coomassie Blue
PSRPPSFSPCPAEDLFPGHRYDGGLDSGFHSVDSGSKRWSGNESTDEFSELSFRISELAREPRGPRERKEDGSADGDPVQIDFIDSHVPGEDEERGTVEEQRPPEL
RUO
LRCH4
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
4034
LRCH4 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ40101/FLJ46315/LRN/LRRN1/LRRN4/PP14183/SAP25
LRCH4
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers