Learn More
Abnova™ Human LOXL4 Partial ORF (NP_115587, 657 a.a. - 755 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00084171-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the lysyl oxidase gene family. The prototypic member of the family is essential to the biogenesis of connective tissue, encoding an extracellular copper-dependent amine oxidase that catalyses the first step in the formation of crosslinks in collagens and elastin. A highly conserved amino acid sequence at the C-terminus end appears to be sufficient for amine oxidase activity, suggesting that each family member may retain this function. The N-terminus is poorly conserved and may impart additional roles in developmental regulation, senescence, tumor suppression, cell growth control, and chemotaxis to each member of the family. [provided by RefSeq]
Sequence: ACANFGEQGVTVGCWDTYRHDIDCQWVDITDVGPGNYIFQVIVNPHYEVAESDFSNNMLQCRCKYDGHRVWLHNCHTGNSYPANAELSLEQEQRLRNNLSpecifications
NP_115587 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
ACANFGEQGVTVGCWDTYRHDIDCQWVDITDVGPGNYIFQVIVNPHYEVAESDFSNNMLQCRCKYDGHRVWLHNCHTGNSYPANAELSLEQEQRLRNNL | |
RUO | |
LOXL4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
84171 | |
LOXL4 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ21889/LOXC | |
LOXL4 | |
Recombinant | |
wheat germ expression system |