Learn More
Abnova™ Human LOXL3 Partial ORF (NP_115992, 171 a.a. - 270 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00084695-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the lysyl oxidase gene family. The prototypic member of the family is essential to the biogenesis of connective tissue, encoding an extracellular copper-dependent amine oxidase that catalyses the first step in the formation of crosslinks in collagens and elastin. A highly conserved amino acid sequence at the C-terminus end appears to be sufficient for amine oxidase activity, suggesting that each family member may retain this function. The N-terminus is poorly conserved and may impart additional roles in developmental regulation, senescence, tumor suppression, cell growth control, and chemotaxis to each member of the family. Alternatively spliced transcript variants of this gene have been reported but their full-length nature has not been determined. [provided by RefSeq]
Sequence: IRPAVGWGRRPLPVTEGLVEVRLPDGWSQVCDKGWSAHNSHVVCGMLGFPSEKRVNAAFYRLLAQRQQHSFGLHGVACVGTEAHLSLCSLEFYRANDTARSpecifications
NP_115992 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
IRPAVGWGRRPLPVTEGLVEVRLPDGWSQVCDKGWSAHNSHVVCGMLGFPSEKRVNAAFYRLLAQRQQHSFGLHGVACVGTEAHLSLCSLEFYRANDTAR | |
RUO | |
LOXL3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
84695 | |
LOXL3 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
LOXL | |
LOXL3 | |
Recombinant | |
wheat germ expression system |